Cole-Parmer®, CBH-400 Recirculator
Leverandør: Cole Parmer
13042-03
STUA13042-03EA 0 NOK
STUA13042-03
Cole-Parmer®, CBH-400 Recirculator
Termostatbad Termostatbad, lukket Termostatbad med kjøling
CBH-400 recirculator offers full-range cooling for quick ramp up and cool-down - saves valuable time.
  • Robust cooling and heating from −10 to 80 °C
  • Compact design maximises bench space and provides quick temperature changes
  • Internal tank design minimizes fluid evaporation
  • 17 LPM recirculator
  • 250 Watt cooling capacity
  • 230 VAC operation
  • 3 year warranty
True thermostatic control laboratory chiller. Removes or adds temperature as required by your application. This closed laboratory chiller is ideal for low load applications such as cooling smallerMore Product Information