Cole-Parmer® Essentials Bottle Top Dispensers with Recirculation Valve
Leverandør: Cole Parmer
25208-31
STUA25208-31EA 0 NOK
STUA25208-31
Cole-Parmer® Essentials Bottle Top Dispensers with Recirculation Valve
Flasketoppdispensere
Save money by conserving reagents.
  • Recirculation valve prevents loss of reagent during priming and purging
  • Smooth plunger movement provides bubble-free dispensing
  • Adjustable nozzle allows users to change the dispensing angle
  • Wetted materials: PTFE, ETP, borosilicate glass, PP, PFA, FEP
  • Autoclavable
These bottle top dispensers conserve reagents with a built-in recirculation valve, allowing users to prime and purge without waste. The PTFE piston with ETP O-ring provide frictionless movement, reducing the occurrence of bubbles in dispenses. TheMore Product Information