Superspher® RP18-E, HPLC Columns
Leverandør: Avantor
SUPERRP18E412FEA 11825 NOK
SUPERRP18E412F
Superspher® RP18-E, HPLC Columns
Kromatografikolonner
A range of 4 µm materials has been marketed under the Superspher® tradename in Europe.
  • Spherical porous silica
  • Base deactivated LiChrospher RP-select B
  • Endcapping options for RP-8 and RP-18
  • Hichrom high efficiency packed columns