TraceCERT®, Supelco® Bromide Standard for Ion Chromatography (IC)
Leverandør: Merck
43147-100ML
SUPL43147-100MLEA 1000 NOK
SUPL43147-100ML
TraceCERT®, Supelco® Bromide Standard for Ion Chromatography (IC)
Standarder Anionstandarder Bromide Standards
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM.