Aluminium 1.000 mg/L in fortynnet salpetersyre (fra Al(NO3)3.9H2O) TraceCERT® enkeltgrunnstoffsstandard til ICP, Supelco®
Leverandør: Merck
61935-100ML-F
SUPL61935-100ML-FEA 1475 NOK
SUPL61935-100ML-F
Aluminium 1.000 mg/L in fortynnet salpetersyre (fra Al(NO3)3.9H2O) TraceCERT® enkeltgrunnstoffsstandard til ICP, Supelco®
Standarder Kationstandarder Aluminium Standards
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST. Certified content incl. uncertainty and expiry date are given on the certificate.