Natriumklorid, Supelco®

Leverandør: Merck

71387-25G
SUPL71387-25GEA 2325 NOK
SUPL71387-25G
Natriumklorid, Supelco®
Natriumklorid
Formel: NaCl
Kokepunkt: 1413 °C (1013 hPa)
Smeltepunkt: 801 °C
Tetthet: 2,16 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003477
CAS-nummer: 7647-14-5
EINECS: 231-598-3
Merck Indeks: 14,08599

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR