Alcometer, recalibration
Leverandør: SURESCREEN
AMCAL
SUREAMCALEA 1800 NOK
SUREAMCAL
Alcometer, recalibration
Narkotester
After 500 uses the alcometer requires recalibration to ensure that the instrument is reading correctly. It is returned to manufacturer for this service, and turn around is five working days from receipt.