Drug testing kit for surfaces
Leverandør: SURESCREEN
PLAB4DS24300_U
SUREPLAB4DS24300_UEA 0 NOK
SUREPLAB4DS24300_U
Drug testing kit for surfaces
Narkotester
Using the swab and buffer, this kit allows swabbing of suspect surfaces or powders to determine if they are amphetamines.
  • Testing drug paraphernalia, surfaces or suspected raw drug
  • Highly accurate and easy to use
  • Results within five minutes
Suitable for customs, security, or anywhere where drug use is a concern.