Trypan blue 0,4% PBS

Leverandør: HyClone products (Cytiva)

Synonymer: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)

SV30084.01
SV30084.01EA 300 NOK
SV30084.01
Trypan blue 0,4% PBS
Trypan blue

Trypan Blue solution, 0,4%, is routinely used as a cell stain to assess cell viability using the dye exclusion test.

Formel: C₃₄H₂₄N₆Na₄O₁₄S₄
MW: 960.82 g/mol
MDL nummer: MFCD00003969
CAS-nummer: 72-57-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR