Surgical blade, No. 3, stitch cutter
Leverandør: SWANN MORTON
0326
SWAN0326EA 1425 NOK
SWAN0326
Surgical blade, No. 3, stitch cutter
Skalpellblad og skaft Skalpellblad
The stainless steel No. 3 or 'baby' stitch cutter is used in conjunction with the No. 3 surgical handle.
  • Convenient and cost effective
  • Sterile
Enables a more aseptic techinique by operating further away from the actual wound which also allows better visibility.
Fits handles 3 Graduated, 3L, 5B, 7, 9, B3 and B3L.