Stitch cutter, disposable, 0526
Leverandør: SWANN MORTON
0526
SWAN0526EA 310 NOK
SWAN0526
Stitch cutter, disposable, 0526
Skalpellblad og skaft Skalpeller
Sterile stitch cutter mounted on a plastic handle allowing extra dexterity and distance from the wound.
  • Stainless steel blade
  • PS handle
Used for pain free removal of modern day suture materials on healing of the wound.