Handle for craft tool
Leverandør: SWANN MORTON
1235
SWAN1235EA 680 NOK
SWAN1235
Handle for craft tool
Skalpellblad og skaft Skalpellskaft
Handle made from impact resistant PS.
  • A thumbscrew is used for blade changing and retaining
  • For use with blades No. 1, 2 or 3
This craft tool is an ideal tool for marquetry, modelling, craft, studio work and horticulture.
Bestillingsinformasjon: Handle only. Blades need to be ordered separately.