Scalpel handle, Cygnetic®
Leverandør: SWANN MORTON
5398
SWAN5398EA 2375 NOK
SWAN5398
Scalpel handle, Cygnetic®
Skalpellblad og skaft Skalpellskaft
This stainless steel handle is designed to assist the OR nurse with safe and easy blade attachment and removal before, during and after the procedure.
  • Safe and easy blade attachment and removal
  • Simple disassembly for cleaning
  • Great strength and stability
  • Handle is non sterile, blades are sterile
Ideal for use within rigorous Orthopaedic and general surgical procedures.
Forpakning: Handle is supplied individually sleeved. Blades (to be ordered separately) are supplied individually V.P.I. protected in metal foil packets.