Gel documentation system, Syngene G:BOX Chemi XX9
Leverandør: Syngene (a division of the Synoptics group)
GBOX-CHEMI-XX9
SYNOGBOX-CHEMI-XX9EA 0 NOK
SYNOGBOX-CHEMI-XX9
Gel documentation system, Syngene G:BOX Chemi XX9
Fotodokumentasjonssystemer
The Syngene G:BOX Chemi XX9 gel documentation system offers optimum performance. Highly sensitive, it is designed for advanced chemiluminescent Western blots, multiplex gels, IR and routine imaging of fluorescent and visible gels and blots. The higher spatial resolution of this system makes it suitable for advanced applications, including 2D. The G:BOX Chemi XX9 includes a super-high resolution, 9 megapixel camera with quantum efficiency >73% at 425 nm, giving outstanding sensitivity for chemiluminescence. Functional control of the G:BOX Chemi XT4 is handled by the GeneSys automaticMore Product Information