Gel documentation system, Syngene G:BOX-F³
Leverandør: Syngene (a division of the Synoptics group)
GBOX-F3-LFB
SYNOGBOX-F3-LFBEA 0 NOK
SYNOGBOX-F3-LFB
Gel documentation system, Syngene G:BOX-F³
Fotodokumentasjonssystemer
The Syngene G:BOX-F³ gel imaging system is designed for general fluorescence applications, including DNA and protein analysis. The range of applications available can be extended by adding lighting modules*. Driven from a database containing hundreds of application protocols, functional control of the G:BOX-F³ is handled by the GeneSys automatic control software, although full manual control is also possible. It has a 3,8 megapixel camera with motorised (f/1,2) lens , which gives high quality images with undetectable noise.
  • CCD camera with 3,8 megapixel resolution and motor driven zoom