Blue light LED transilluminators, Syngene UltraSlim
Leverandør: Syngene (a division of the Synoptics group)
GEULTRASLIMLED
SYNOGEULTRASLIMLEDEA 9850 NOK
SYNOGEULTRASLIMLED
Blue light LED transilluminators, Syngene UltraSlim
Transilluminatorer Blått lys transilluminatorer
UltraSlim-LED transilluminators for superior gel illumination.
  • Compact
  • Even blue epi LED illumination - high sensitivity with improved S/N ratio
  • Image a range of fluorophores, eg. EtBr, SYBR®Safe, GelGreen, SYBR®Gold, SYBR®Green, SYPRO®Ruby, SYPRO®Orange, GelRed™
  • LED is safe to skin and eyes with no damage to DNA