Chemiluminescence imaging systems, Syngene GeneGnome XRQ
Leverandør: Syngene (a division of the Synoptics group)
GGNOME-XRQ-NPC
SYNOGGNOME-XRQ-NPCEA 0 NOK
SYNOGGNOME-XRQ-NPC
Chemiluminescence imaging systems, Syngene GeneGnome XRQ
Fotodokumentasjonssystemer
The Syngene GeneGnome XRQ is dedicated to chemiluminescence imaging. The system is built for high performance and automation and features a next generation high quantum efficiency CCD camera for even greater sensitivity. A very simple set-up process means a single click will automatically capture a quality image of any Western blot.
  • Configured for maximum sensitivity to ensure even the faintest band on a blot can be captured
  • High quantum efficiency cooled CCD camera (73% @ 425 nm) is very sensitive to low level light emissions from a blot
  • Peltier cooled