Fenol rødt, BAKER ANALYZED® ACS indicator, J.T.Baker®

Leverandør: Avantor

Synonymer: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

T254-01EA 1425 NOK
T254-01
Fenol rødt, BAKER ANALYZED® ACS indicator, J.T.Baker®
Fenol rødt
Formel: C₁₉H₁₄O₅S
MW: 354.38 g/mol
Smeltepunkt: >300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003552
CAS-nummer: 143-74-8
EINECS: 205-609-7
Merck Indeks: 13,07329

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements Passes Test
For Laboratory,Research,or Manufacturing Use Passes Test
Clarity of Solution Passes Test
Visual Transition Interval - pH…6.8 (Yellow) Passes Test
Visual Transition Interval - pH…8.2 (Red) Passes Test
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR