Phenol red sodium salt, powder, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor
Warning

Synonymer: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide

T265-01EA 2575 NOK
T265-01
Phenol red sodium salt, powder, BAKER ANALYZED® ACS, J.T.Baker®
Phenol red sodium salt

Powder. Lot analysis on label.

Formel: C₁₉H₁₃NaO₅S
MW: 376.35 g/mol
Smeltepunkt: 285
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00066901
CAS-nummer: 34487-61-1
EINECS: 252-057-8
Merck Indeks: 12,07397

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements Passes Test
For Laboratory,Research,or Manufacturing Use Passes Test
ACS - Clarity of Solution Passes Test
Visual Transition Interval - pH…8.2 (Red) Passes Test
Visual Transition Interval - pH…6.8 (Yellow) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR