Toluidinblått O, technical, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: TBO, 3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chloride, 3-Amino-7-(dimethylamino)-2-methylphenothiazin-5-ium chloride, Tolonium chloride, Basic Blue 17, Blutene chloride

MQ200

T3260-100G T3260-25G T3260-5G
T3260-100GEA 13890 NOK
T3260-100G T3260-25G T3260-5G
Toluidinblått O, technical, Sigma-Aldrich®
Toluidinblått O

Toluidine Blue O (TBO) is a thiazine dye of the quinone-imine family and is cationic in nature. It has been employed in polychromatic staining of paraffin embedded plant cell walls. It is a metachromatic dye suitable for a wide variety of histological staining procedures. A metachromatic dye suitable for a wide variety of histological staining procedures. Toluidine Blue O has been used to stain a variety of specimens like mesenchymal cells, taste buds, osteoblasts, matrix, and more.

Formel: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
Lagringstemperatur: Room Temperature
MDL nummer: MFCD00011934
CAS-nummer: 92-31-9
EINECS: 202-146-2
Merck Indeks: 12,09658
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b68e54e7aa7ac291c3a6c1e795308f18da86b2c&vid=60380f47bd5001b6d4c2dcdf8e15df28ba66eedf&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Solubility (Colour), c = 4 mg/4 ml; Water Dark Blue to Very Dark Blue and Dark Purple to Very Dark Purple and Blue-Purple
Carbon 47.1 - 61.9 %
Nitrogen 11.0 - 14.4 %
Dye Content Approximately 85%

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR