Blå tetrazoliumklorid for colorimetric determination of reducing compounds, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: Blue Tetrazolium, 3,3′-(3,3′-Dimethoxy[1,1′-biphenyl]-4,4′-diyl)-bis(2,5-diphenyl-2H-tetrazolium) dichloride, 3,3'-(3,3'-Dimethoxy-4,4'-biphenylene)bis(2,5-diphenyl-2H-tetrazolium chloride), BTC, 3,3′-Dianisole-4,4′-bis(3,5-diphenyltetrazolium chloride), Blue Tetrazolium chloride

M-Clarity™ quality level = MQ200

T4375-10G T4375-1G T4375-25G T4375-5G
T4375-10GEA 3530 NOK
T4375-10G T4375-1G T4375-25G T4375-5G
Blå tetrazoliumklorid for colorimetric determination of reducing compounds, Sigma-Aldrich®
Blå tetrazoliumklorid

Tetrazolium blue chloride (BTC) is a colorless tetrazolium compound that upon reduction forms a colored diformazan. The reactivity has been applied to the analysis of reducing compounds including the nanomolar detection of reducing sugars and carbidopa and methyldopa. BTC has been used for histological staining of mouse coronal brain sections and rat brain sections. In combination with 4-nitrocatechol, tetrazolium blue chloride forms a ternary complex with molybdenum(VI) for extraction and spectrophotometric determination of molybdenum. It has also been used to stain Francisella tularensis cells for tularemic microagglutination test.

Formel: C₄₀H₃₂Cl₂N₈O₂
MW: 727.65 g/mol
Smeltepunkt: 255 °C (dec., lit.)
Lagringstemperatur: Room Temperature
MDL nummer: MFCD00040933
CAS-nummer: 1871-22-3
EINECS: 217-488-8
Merck Indeks: 12,09380
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=825ba12666da399ee548628826b021d3cbd4ba71&vid=825ba12666da399ee548628826b021d3cbd4ba71&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf42164c85c3ff11937c1db791fff066d2010ddb&vid=2fc65401f1ae23533e1bdd2b18b303813894aa03&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Colour) Yellow
Appearance (Form) Powder
Solubility (Colour) Yellow to Yellow-Green
Solubility (Turbidity)
50 mg/mL, MeOH
Clear
Infrared spectrum Conforms to Structure
Carbon 63.4 - 67.4 %
Nitrogen 14.9 - 15.9 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR