Human cancer cell line blot
Leverandør: G Bioscienes
TB55
TB55EA 43350 NOK
TB55
Human cancer cell line blot
Elektroforese reagenser Western Blotting Reagents
Ready to screen human cancer cell line blots.
These premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4 to 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using Western transfer buffer. The blots are then ready to be blocked and probed with the antibodies of choice.
Features
Blot lane order:
Protein marker
Acute promyelocytic leukemia (HL-60)
T lymphocyte lymphoma (HUT-78)
B cell lymphoma (BJAB)
Mammary adenocarcinoma (MCF7)
Amelanotic melanoma (C32)
Acute T cell leukemia (JURKAT)
Chronic Myelogenous leukemia (K562)
Epidermoid carcinoma (A-431)
Epithlioid carcinoma (HeLa)
Burkitt's lymphoma (Ramos)
Prostate adenocarcinoma (LNCap)
Hepatoblastoma (HepG2)
Neuroepithelioma (SK-N-MC)