Acetaldehyde oxime (mixture of syn and anti) ≥99.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Acetaldoxime

A0006-25ML
TCIAA0006-25MLEA 515 NOK
TCIAA0006-25ML
Acetaldehyde oxime (mixture of syn and anti) ≥99.0% (by GC)
Acetaldehyde oxime (mixture of syn and anti)
Formel: C₂H₅NO
MW: 59.07 g/mol
Kokepunkt: 115°C
MDL nummer: MFCD00002124
CAS-nummer: 107-29-9
EINECS: 203-479-6
FN: 2332
ADR: 3 III
Merck Indeks: 14,00042

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 99.0 % (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR