Vinylacetat monomer ≥99.0% (by GC) stabilised

Leverandør: TCI
Danger

Synonymer: Acetic acid vinyl ester

Stabilisering: stabilisert med hydroquinon

A0045-500ML
TCIAA0045-500MLEA 245 NOK
TCIAA0045-500ML
Vinylacetat monomer ≥99.0% (by GC) stabilised
Vinylacetat
Formel: C₄H₆O₂
MW: 86,09 g/mol
Kokepunkt: 72°C
Flash punkt: -8°C
MDL nummer: MFCD00008713
CAS-nummer: 108-05-4
EINECS: 203-545-4
FN: 1301
ADR: 3,II
Merck Indeks: 14,09992

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR