N-Acetylglycine(Ac-Gly-OH) ≥99.0% (by titrimetric analysis)

Leverandør: TCI

A0093-25G A0093-500G A0093-100G
TCIAA0093-25GEA 220 NOK
TCIAA0093-25G TCIAA0093-500G TCIAA0093-100G
N-Acetylglycine(Ac-Gly-OH) ≥99.0% (by titrimetric analysis)
N-Acetylglycine(Ac-Gly-OH)
Formel: C₄H₇NO₃
MDL nummer: MFCD00004275
CAS-nummer: 543-24-8
EINECS: 208-839-6
Merck Indeks: 14,00080
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ed83a21dc727fc21a98ef0a3d592e39f8dbbfb5&vid=9ed83a21dc727fc21a98ef0a3d592e39f8dbbfb5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 99.0 %
Solubility in Water almost transparency
Drying loss max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR