trans-Aconitic acid 98,0 HPLC, T

Leverandør: TCI
Warning

Synonymer: Trans-Propene-1,2,3-Tricarboxylic Acid, (E)-Prop-1-ene-1,2,3-tricarboxylic acid

A0127-25G A0127-500G
TCIAA0127-25GEA 675 NOK
TCIAA0127-25G TCIAA0127-500G
trans-Aconitic acid 98,0 HPLC, T
trans-Aconitic acid
Formel: C₆H₆O₆
MW: 174.11 g/mol
MDL nummer: MFCD00002721
CAS-nummer: 4023-65-8
EINECS: 223-688-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be887b8b6ec29c6b910ab11c3a91b0b411b0ef5c&vid=be887b8b6ec29c6b910ab11c3a91b0b411b0ef5c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR