ɑ-Amino-γ-butyrolactone hydrobromide 98,0 N, T

Leverandør: TCI
Warning

Synonymer: DL-Homoserine lactone hydrobromide

A0283-1G A0283-5G
TCIAA0283-1GEA 515 NOK
TCIAA0283-1G TCIAA0283-5G
ɑ-Amino-γ-butyrolactone hydrobromide 98,0 N, T
alpha-Amino-gamma-butyrolactone hydrobromide
Formel: C₄H₇NO₂
MW: 182.02 g/mol
MDL nummer: MFCD00012723
CAS-nummer: 6305-38-0
EINECS: 228-614-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR