Auramine O ≥80.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

A0334-25G
TCIAA0334-25GEA 365 NOK
TCIAA0334-25G
Auramine O ≥80.0% (by HPLC, titration analysis)
Auramine O
Formel: C₁₇H₂₂ClN₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012484
CAS-nummer: 2465-27-2
EINECS: 219-567-2
FN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=255e6ab1af21bc764fd2448d6cf8b23c3e1704a2&vid=255e6ab1af21bc764fd2448d6cf8b23c3e1704a2&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Amber to Dark green powder to crystaline
Purity (HPLC) min. 80.0 area%
Purity (Argentmetric Titration) min. 80.0 %
Lambda max. 430.0 to 434.0 nm (H2O)
Absorbance (E1%1cm) min. 900 (430.0 to 434.0 nm, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR