2-Amino-5-mercapto-1,3,4-thiadiazole ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 5-Amino-1,3,4-thiadiazole-2-thiol

A0425-25G A0425-500G
TCIAA0425-25GEA 270 NOK
TCIAA0425-25G TCIAA0425-500G
2-Amino-5-mercapto-1,3,4-thiadiazole ≥98.0% (by HPLC, titration analysis)
2-Amino-5-mercapto-1,3,4-thiadiazole
Formel: C₂H₃N₃S₂
Smeltepunkt: 234°C
MDL nummer: MFCD00003108
CAS-nummer: 2349-67-9
EINECS: 219-078-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef571913ad3be1cd236d03b8339361e7c7f8e742&vid=ef571913ad3be1cd236d03b8339361e7c7f8e742&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in 1mol/L NaOH faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR