Nickel(II) 2-amino-5-methylbenzenesulphonate ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: 2-Amino-5-methylbenzenesulphonic Ac, 4-Aminotoluene-3-sulphonic acid Nickel(II) Salt, 4-Methylaniline-2-sulphonic acid Nickel(II) Salt, Nickel(II) 4-Methylaniline-2-sulphonate, p-Toluidine-2-sulphonic acid Nickel(II) Salt, Nickel(II) 4-Aminotoluene-3-sulphonate

A0430-25G
TCIAA0430-25GEA 1700 NOK
TCIAA0430-25G
Nickel(II) 2-amino-5-methylbenzenesulphonate ≥95.0% (by titrimetric analysis)
Nickel(II) 2-amino-5-methylbenzenesulphonate
Formel: C₇H₉NO₃S
MW: 431,10 g/mol
MDL nummer: MFCD00135316
CAS-nummer: 375387-13-6

Bestill nå

Spesifikasjon Testresultater

Appearance Green to Dark green White to Light yellow
Purity (Chelometric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR