α,α,α-Trifluoro-o-toluidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

A0433-25ML A0433-500ML
TCIAA0433-25MLEA 220 NOK
TCIAA0433-25ML TCIAA0433-500ML
α,α,α-Trifluoro-o-toluidine ≥98.0% (by GC, titration analysis)
α,α,α-Trifluoro-o-toluidine
Formel: C₇H₆F₃N
Kokepunkt: 171°C
Smeltepunkt: –33 °C
Tetthet: 1,282 g/cm³ (20 °C)
Flash punkt: 55°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007718
CAS-nummer: 88-17-5
EINECS: 201-806-7
FN: 2942
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66193824ad5b87ed5eb8b2676243585858dfe2ea&vid=66193824ad5b87ed5eb8b2676243585858dfe2ea&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid to slightly cloudy liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR