Azure II

Leverandør: TCI
Warning

Synonymer: C.I. 52010/52015, 3,7 Bis(dimethylamino)phenothiazin-5-ium chloride and 3-(Dimethylamino)-7-(methylamino)-phenothiazin-5-ium chloride, Azur B/Methylene Blue 1:1, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride mixt. with 3-(dimethylamino)-7-(methylamino)phenothiazin-5-ium chloride(1:1), Methylene Azure II

A0574-1G A0574-25G
TCIAA0574-1GEA 220 NOK
TCIAA0574-1G TCIAA0574-25G
Azure II
Azure II
Formel: C₁₆H₁₈N₃S·C₁₅H₁₆N₃S·2Cl
MW: 625.68 g/mol
MDL nummer: MFCD00080716
CAS-nummer: 37247-10-2
Merck Indeks: 14,06058

Bestill nå

Spesifikasjon Testresultater

Appearance Green to Dark green powder to crystal
Lambda max. 650.0 to 664.0 nm (H2O)
Absorbance (E1%1cm) min. 1500 (H2O, 650.0 to 664.0 nm) (calcd.on dried substance)
Drying loss max. 10.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR