Acid red 27 ≥90.0% (by UV/Vis)

Leverandør: TCI

Synonymer: Amaranth, Sodium 3-hydroxy-4-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-2,7-disulphonate, Bordeaux S, Food Red No. 2, Azo Rubine S, Wool Red 40 F, Naphthol Red

A0583-25G
TCIAA0583-25GEA 340 NOK
TCIAA0583-25G
Acid red 27 ≥90.0% (by UV/Vis)
Acid red 27
Formel: C₂₀H₁₄N₂O₁₀S₃
Smeltepunkt: 220°C
MDL nummer: MFCD00004076
CAS-nummer: 915-67-3
EINECS: 213-022-2
Merck Indeks: 14,00375

Bestill nå

Spesifikasjon Testresultater

Appearance Red to Dark red to Brown powder to crystal
Purity (UV-vis method) min. 90.0 % (after drying)
Drying loss max. 5.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR