Acid red 1

Leverandør: TCI

Synonymer: Red 2G, C.I. 18050, Amido Naphthol Red G, Sodium 5-acetamido-4-hydroxy-3-(phenyldiazenyl)naphthalene-2,7-disulphonate, 5-Acetamido-4-hydroxy-3-phenylazo-2,7-naphthalene-disulphonic acid disodium salt, Pontacyl Carmine, Azophloxine

A0585-25G A0585-500G
TCIAA0585-25GEA 310 NOK
TCIAA0585-25G TCIAA0585-500G
Acid red 1
Acid red 1
Formel: C₁₈H₁₅N₃O₈S₂
MDL nummer: MFCD00003954
CAS-nummer: 3734-67-6
EINECS: 223-098-9

Bestill nå

Spesifikasjon Testresultater

Appearance Red to Dark red to Brown powder to crystal
Lambda max. 503.0 to 507.0 nm (H2O)
Lambda max.2 530.0 to 534.0 nm (H2O)
Absorbance (E1%1cm) min. 250 (H2O, 503.0 to 507.0 nm)
Absorbance (E1%1cm)2 min. 250 (H2O, 530.0 to 534.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR