ACES 99,0 Titrimetric Good's buffer comp. for biological resea

Leverandør: TCI

Synonymer: ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid

A0700-25G
TCIAA0700-25GEA 595 NOK
TCIAA0700-25G
ACES 99,0 Titrimetric Good's buffer comp. for biological resea
ACES
Formel: H₂NCOCH₂NHCH₂CH₂SO₃H
MW: 182.2 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008030
CAS-nummer: 7365-82-4
EINECS: 230-908-4
Merck Indeks: 13,00037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfe9b43a765df2751dc5467423db84035cc52aa6&vid=f5d08bbc6574e75163bc53bf993c60aec9b97c0f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White powder to crystal
Purity (Neutralization titration) min. 99.0 %
Absorbance max. 0.015 (0.1mol/L、H2O、260nm)
Solubility in hot Water almost transparency
Iron max. 5 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR