4-Chloro-o-toluidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

A0704-25G A0704-500G A0704-100G
TCIAA0704-25GEA 525 NOK
TCIAA0704-25G TCIAA0704-500G TCIAA0704-100G
4-Chloro-o-toluidine ≥98.0% (by GC, titration analysis)
4-Chloro-o-toluidine
Formel: C₇H₈ClN
Kokepunkt: 241°C
Tetthet: 1,14 g/cm³ (20 °C)
MDL nummer: MFCD00007842
CAS-nummer: 95-69-2
EINECS: 202-441-6
FN: 2239
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0c6e7f6cc985ac485c508a5eaea6d659bb8bfd4&vid=f0c6e7f6cc985ac485c508a5eaea6d659bb8bfd4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR