Trimethyl-trans-aconitate 97,0 GC_ASSAY_METHOD

Leverandør: TCI
Warning

Synonymer: trans-Aconitic Acid Trimethyl Ester

A0725-25G A0725-5G
TCIAA0725-25GEA 3600 NOK
TCIAA0725-25G TCIAA0725-5G
Trimethyl-trans-aconitate 97,0 GC_ASSAY_METHOD
Trimethyl-trans-aconitate
Formel: C₉H₁₂O₆
MW: 216.19 g/mol
Kokepunkt: 160°C
MDL nummer: MFCD00059304
CAS-nummer: 4271-99-2

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR