Acetophenone oxime ≥98.0% (by GC, total nitrogen)

Leverandør: TCI
Warning

Synonymer: Methyl phenyl ketoxime, 1-Phenyl-1-ethanone oxime, Acetophenone Oxime

A0807-25G
TCIAA0807-25GEA 650 NOK
TCIAA0807-25G
Acetophenone oxime ≥98.0% (by GC, total nitrogen)
Acetophenone oxime
Formel: C₈H₉NO
MW: 135,17 g/mol
Kokepunkt: 119°C
Smeltepunkt: 60 °C
MDL nummer: MFCD00013931
CAS-nummer: 613-91-2
EINECS: 210-360-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d133f3d233a58d2cda13e162cf529e20f2ab186f&vid=d133f3d233a58d2cda13e162cf529e20f2ab186f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 58.0 to 62.0 deg-C
Solubility in EtOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR