cis-3-Hexenyl acetate ≥97.0% (by GC)

Leverandør: TCI
Warning

A0888-25ML
TCIAA0888-25MLEA 350 NOK
TCIAA0888-25ML
cis-3-Hexenyl acetate ≥97.0% (by GC)
cis-3-Hexenyl acetate
Formel: C₈H₁₄O₂
MW: 142,20 g/mol
Kokepunkt: 169°C
Flash punkt: 57°C
MDL nummer: MFCD00036563
CAS-nummer: 3681-71-8
EINECS: 222-960-1
FN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=512c02c453e35e8f1c1efcdc0152d04a63faec97&vid=512c02c453e35e8f1c1efcdc0152d04a63faec97&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR