Amitriptyline hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

Synonymer: Elavil hydrochloride, 5-[3-(Dimethylamino)propylidene]dibenzosuberane hydrochloride, Amitriptyline Hydrochloride, 3-(10,11-Dihydro-5H-dibenzo[a,d]cycloheptene-5-ylidene)-N,N-dimethyl-1-propanamine hydrochloride, Endep hydrochloride, Levate hydrochloride

A0908-25G
TCIAA0908-25GEA 1100 NOK
TCIAA0908-25G
Amitriptyline hydrochloride ≥98.0% (by HPLC, titration analysis)
Amitriptyline hydrochloride
Formel: C₂₀H₂₃N
MDL nummer: MFCD00012537
CAS-nummer: 549-18-8
EINECS: 208-964-6
FN: 2811
ADR: 6.1,III
Merck Indeks: 14,00487

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 195.0 to 199.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR