3-Methyl-2-butenyl acetate ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Acetic Acid Prenyl Ester, Acetic Acid 3-Methyl-2-butenyl Ester, 3-Methylbut-2-en-1-yl acetate, Prenyl Acetate

A1148-25ML
TCIAA1148-25MLEA 220 NOK
TCIAA1148-25ML
3-Methyl-2-butenyl acetate ≥98.0% (by GC)
3-Methyl-2-butenyl acetate
Formel: C₇H₁₂O₂
MW: 128.17 g/mol
Kokepunkt: 128°C
MDL nummer: MFCD00036569
CAS-nummer: 1191-16-8
EINECS: 214-730-4
FN: 3272
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52334240eea76410d236334ed00dbead1998f7be&vid=52334240eea76410d236334ed00dbead1998f7be&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR