Acid red 13

Leverandør: TCI

Synonymer: Fast Red E

A1243-10G
TCIAA1243-10GEA 700 NOK
TCIAA1243-10G
Acid red 13
Acid red 13
Formel: C₂₀H₁₄N₂O₇S₂
MDL nummer: MFCD00191366
CAS-nummer: 2302-96-7
EINECS: 218-958-5

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown to Dark purple powder to crystal
Lambda max. 503.0 to 507.0 nm (0.02mol/L AcONH4 sol.)
Absorbance (E1%1cm) min. 350 (0.02mol/L AcONH4 sol., 503.0 to 507.0 nm)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR