1-Amino-2-methyl-9,10-anthraquinone ≥90.0% (by total nitrogen basis)

Leverandør: TCI

A1255-25G
TCIAA1255-25GEA 675 NOK
TCIAA1255-25G
1-Amino-2-methyl-9,10-anthraquinone ≥90.0% (by total nitrogen basis)
1-Amino-2-methyl-9,10-anthraquinone
Formel: C₁₅H₁₁NO₂
MDL nummer: MFCD00001220
CAS-nummer: 82-28-0
EINECS: 201-408-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22e3d440e0a415db12a18b02f6bee7843e07ff89&vid=22e3d440e0a415db12a18b02f6bee7843e07ff89&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Amber to Dark red powder to crystal
Purity (HPLC) min. 85.0 area%
Purity (with Total Nitrogen) min. 90.0 %
Melting point 198.0 to 204.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR