Amantadine sulphate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

A1319-25G
TCIAA1319-25GEA 540 NOK
TCIAA1319-25G
Amantadine sulphate ≥98.0% (by titrimetric analysis)
Amantadine sulphate
Formel: C₁₀H₁₉NO₄S
Smeltepunkt: 299…301 °C
MDL nummer: MFCD00077251
CAS-nummer: 31377-23-8
EINECS: 250-604-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87a66afd99b281f9d5057d9e79b018f5a387b9f3&vid=87a66afd99b281f9d5057d9e79b018f5a387b9f3&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR