2-Chloro-p-toluidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

A1437-25G A1437-5G
TCIAA1437-25GEA 1200 NOK
TCIAA1437-25G TCIAA1437-5G
2-Chloro-p-toluidine ≥98.0% (by GC, titration analysis)
2-Chloro-p-toluidine
Formel: C₇H₈ClN
Kokepunkt: 224°C
Smeltepunkt: 7°C
Tetthet: 1,151 g/cm³ (20 °C)
Flash punkt: 99°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007666
CAS-nummer: 615-65-6
EINECS: 210-440-7
FN: 3429
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ebd07836ee72782d6cdb91a3262b7c8c3590a3d&vid=5ebd07836ee72782d6cdb91a3262b7c8c3590a3d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR