cis-4-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI

A1448-1G A1448-25G A1448-5G
TCIAA1448-1GEA 435 NOK
TCIAA1448-1G TCIAA1448-25G TCIAA1448-5G
cis-4-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
cis-4-Aminocyclohexanecarboxylic acid
Formel: C₇H₁₃NO₂
MDL nummer: MFCD00191730
CAS-nummer: 3685-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5af8e4ddd105349cc2ebb50cf90557a7a8ad6d0&vid=f5af8e4ddd105349cc2ebb50cf90557a7a8ad6d0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR