Kloramben ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 3-Amino-2,5-dichlorobenzoic acid, Chloramben

A1537-5G
TCIAA1537-5GEA 1925 NOK
TCIAA1537-5G
Kloramben ≥98.0% (by titrimetric analysis)
Kloramben
Formel: C₇H₅Cl₂NO₂
Kokepunkt: 349 °C (1013 hPa)
Smeltepunkt: 200…201 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00065093
CAS-nummer: 133-90-4
EINECS: 205-123-5
Merck Indeks: 12,02115
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=391f385013c3a10589030fb9502c3e21af0a3bdc&vid=391f385013c3a10589030fb9502c3e21af0a3bdc&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR