4-Chloro-m-toluidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 4-Chloro-3-methylaniline, 5-Amino-2-chlorotoluene

A1610-5G
TCIAA1610-5GEA 570 NOK
TCIAA1610-5G
4-Chloro-m-toluidine ≥98.0% (by GC, titration analysis)
4-Chloro-m-toluidine
Formel: C₇H₈ClN
Kokepunkt: 241 °C (1013 hPa)
Smeltepunkt: 83…84 °C
MDL nummer: MFCD00066332
CAS-nummer: 7149-75-9
FN: 2239
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6f0e5fe73d259bf48c37c76939fe129f3e8fcd3&vid=b6f0e5fe73d259bf48c37c76939fe129f3e8fcd3&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 83.0 to 87.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR