Acetaldehyd ∼2% in DMF for detection of primary and secondary amines

Leverandør: TCI
Danger

Synonymer: Ethanal

A1640-10ML
TCIAA1640-10MLEA 435 NOK
TCIAA1640-10ML
Acetaldehyd ∼2% in DMF for detection of primary and secondary amines
Acetaldehyd
Formel: CH₃CHO
MW: 44.05 g/mol
Kokepunkt: 20.1
Smeltepunkt: -123
Flash punkt: -27
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00006991
CAS-nummer: 75-07-0
EINECS: 200-836-8
FN: 1992
ADR: 3 III
Merck Indeks: 14,00039
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5b07aaac7cb312f7b9035077e42772009342904&vid=b4c7c96cdc0b0eedea5780f399f7aec84da0f876&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Concentration (Oxime Formation) 2.0 to 2.3 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR