(S)-(+)-Abscisic acid ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: ABA, (+)-cis,trans ABA, (S)-5-(1-Hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl)-3-methyl-(2Z,4E)-pentadienoic acid, (S)-(+)-Abscisic acid, Dormin, S-ABA, (+)-Abscisic acid

A1698-100MG
TCIAA1698-100MGEA 1375 NOK
TCIAA1698-100MG
(S)-(+)-Abscisic acid ≥98.0% (by HPLC)
(S)-(+)-Abscisic acid
Formel: C₁₅H₂₀O₄
MW: 264,32 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00066545
CAS-nummer: 21293-29-8
EINECS: 244-319-5
Merck Indeks: 14,00011

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 160.0 to 165.0 deg-C
Specific rotation [a]20/D +400.0 to +420.0 deg (C=0.2, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR